N-Boc-D-alaninol, 98%, ee 98%, Thermo Scientific Chemicals
N-Boc-D-alaninol, 98%, ee 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-D-alaninol, 98%, ee 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number H27887.14
also known as H27887-14
Price (USD)/ Each
375.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
375.00
Add to cart
N-Boc-D-alaninol, 98%, ee 98%, Thermo Scientific Chemicals
Catalog numberH27887.14
Price (USD)/ Each
375.00
-
Add to cart
Chemical Identifiers
CAS106391-86-0
IUPAC Nametert-butyl N-[(2R)-1-hydroxypropan-2-yl]carbamate
Molecular FormulaC8H17NO3
InChI KeyPDAFIZPRSXHMCO-ZCFIWIBFSA-N
SMILESC[C@H](CO)NC(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)>97.5%
Enantiomeric excess>98.0% (Supplier's Certificate of Analysis)
Optical Rotation+11? ?1? (c=1 in Chloroform)
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only