Thermo Scientific Chemicals

(2S,3aS,7aS)-Octahydroindole-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: H27894.03
1 g, Each
Thermo Scientific Chemicals

(2S,3aS,7aS)-Octahydroindole-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: H27894.03
1 g, Each
Quantity
Catalog number: H27894.03
also known as H27894-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
80875-98-5
IUPAC Name
octahydro-1H-indole-2-carboxylic acid
Molecular Formula
C9H15NO2
InChI Key
CQYBNXGHMBNGCG-UHFFFAOYNA-N
SMILES
OC(=O)C1CC2CCCCC2N1
Optical Rotation
-50.0 ± 1.0? (C=1 in methanol)
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Powder

Description

An intermediate in the synthesis of Perindopril. The completely diastereoselective α-alkylation of oxazolidinone provides a very convenient and concise route to enantiopure α-tetrasubstituted derivatives of this Oic stereoisomer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An intermediate in the synthesis of Perindopril. The completely diastereoselective α-alkylation of oxazolidinone provides a very convenient and concise route to enantiopure α-tetrasubstituted derivatives of this Oic stereoisomer.

Solubility
Soluble in methanol and water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at -20°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text