6-Chloro-3-pyridineacetic acid, 97%, Thermo Scientific Chemicals
6-Chloro-3-pyridineacetic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Chloro-3-pyridineacetic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number H27966.18
also known as H27966-18
Price (USD)/ Each
270.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
270.00
Add to cart
6-Chloro-3-pyridineacetic acid, 97%, Thermo Scientific Chemicals
Catalog numberH27966.18
Price (USD)/ Each
270.00
-
Add to cart
Chemical Identifiers
CAS39891-13-9
IUPAC Name2-(6-chloropyridin-3-yl)acetic acid
Molecular FormulaC7H6ClNO2
InChI KeyXWVPSJPQWOVRHJ-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=C(Cl)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (HPLC)≥96.0%
Appearance (Color)Pale cream to cream to brown
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only