1,4-Bis(methoxydimethylsilyl)benzene, 97%, Thermo Scientific Chemicals
1,4-Bis(methoxydimethylsilyl)benzene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,4-Bis(methoxydimethylsilyl)benzene, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H28165.06
also known as H28165-06
Price (USD)/ Each
358.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
358.00
Add to cart
1,4-Bis(methoxydimethylsilyl)benzene, 97%, Thermo Scientific Chemicals
Catalog numberH28165.06
Price (USD)/ Each
358.00
-
Add to cart
Chemical Identifiers
CAS131182-68-8
IUPAC Namemethoxy[4-(methoxydimethylsilyl)phenyl]dimethylsilane
Molecular FormulaC12H22O2Si2
InChI KeyVQECZZRUEDMVCU-UHFFFAOYSA-N
SMILESCO[Si](C)(C)C1=CC=C(C=C1)[Si](C)(C)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.4835-1.4885 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only