(R)-(+)-1-Boc-3-aminopyrrolidine, 97%, Thermo Scientific Chemicals
(R)-(+)-1-Boc-3-aminopyrrolidine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(R)-(+)-1-Boc-3-aminopyrrolidine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H28188.06
also known as H28188-06
Price (USD)/ Each
460.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
460.00
Add to cart
(R)-(+)-1-Boc-3-aminopyrrolidine, 97%, Thermo Scientific Chemicals
Catalog numberH28188.06
Price (USD)/ Each
460.00
-
Add to cart
Chemical Identifiers
CAS147081-49-0
IUPAC Nametert-butyl 3-aminopyrrolidine-1-carboxylate
Molecular FormulaC9H18N2O2
InChI KeyCMIBWIAICVBURI-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)N1CCC(N)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Optical Rotation2.8 ± 0.1? (C=1 in CHCl3)
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4665-1.4725 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only