N(epsilon)-Boc-N(alpha)-Fmoc-D-lysine, 98%
N(epsilon)-Boc-N(alpha)-Fmoc-D-lysine, 98%
Thermo Scientific Chemicals

N(epsilon)-Boc-N(alpha)-Fmoc-D-lysine, 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H28301.03
also known as H28301-03
Price (USD)/ Each
64.60
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
64.60
Add to cart
N(epsilon)-Boc-N(alpha)-Fmoc-D-lysine, 98%
Catalog numberH28301.03
Price (USD)/ Each
64.60
-
Add to cart
Chemical Identifiers
CAS92122-45-7
IUPAC Name(2R)-6-{[(tert-butoxy)carbonyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Molecular FormulaC26H32N2O6
InChI KeyUMRUUWFGLGNQLI-ANBDAQEENA-N
SMILESCC(C)(C)OC(=O)NCCCC[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (HPLC)≥97.5%
Optical Rotation10? to 15? (C=1 in DMF)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only