2,6-Dimethoxynaphthalene, 99%, Thermo Scientific Chemicals
2,6-Dimethoxynaphthalene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Dimethoxynaphthalene, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H28366.03
also known as H28366-03
Price (USD)/ Each
65.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
65.00
Add to cart
2,6-Dimethoxynaphthalene, 99%, Thermo Scientific Chemicals
Catalog numberH28366.03
Price (USD)/ Each
65.00
-
Add to cart
Chemical Identifiers
CAS5486-55-5
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Appearance (Color)Pale cream to cream
Melting Point (clear melt)151.0-157.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only