Thermo Scientific Chemicals

(R,R)-DIPAMP, 90%, Thermo Scientific Chemicals

Catalog number: H28374.03
1 g, Each
Thermo Scientific Chemicals

(R,R)-DIPAMP, 90%, Thermo Scientific Chemicals

Catalog number: H28374.03
1 g, Each
Quantity
Catalog number: H28374.03
also known as H28374-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
55739-58-7
IUPAC Name
(2-methoxyphenyl)({2-[(2-methoxyphenyl)(phenyl)phosphanyl]ethyl})phenylphosphane
Molecular Formula
C28H28O2P2
InChI Key
QKZWXPLBVCKXNQ-UHFFFAOYNA-N
SMILES
COC1=C(C=CC=C1)P(CCP(C1=CC=CC=C1)C1=C(OC)C=CC=C1)C1=CC=CC=C1
Appearance (Color)
White
Form
Crystalline powder
Assay (Non-aqueous acid-base Titration)
≤88.0%
Assay by NMR
≥88.0% (31P-NMR)
Comment
Assayed EITHER by titration OR NMR

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text