Chemical Identifiers
CAS55739-58-7
IUPAC Name(2-methoxyphenyl)({2-[(2-methoxyphenyl)(phenyl)phosphanyl]ethyl})phenylphosphane
Molecular FormulaC28H28O2P2
InChI KeyQKZWXPLBVCKXNQ-UHFFFAOYNA-N
SMILESCOC1=C(C=CC=C1)P(CCP(C1=CC=CC=C1)C1=C(OC)C=CC=C1)C1=CC=CC=C1
View more