Dimethylphenylsilanol sodium salt, 97%, Thermo Scientific Chemicals
Dimethylphenylsilanol sodium salt, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethylphenylsilanol sodium salt, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H28457.03
also known as H28457-03
Price (USD)/ Each
122.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
122.00
Add to cart
Dimethylphenylsilanol sodium salt, 97%, Thermo Scientific Chemicals
Catalog numberH28457.03
Price (USD)/ Each
122.00
-
Add to cart
Chemical Identifiers
CAS7646-75-5
IUPAC Namesodium dimethyl(phenyl)silanolate
Molecular FormulaC8H11NaOSi
InChI KeyNPZJKOMUCXNLBN-UHFFFAOYSA-N
SMILES[Na+].C[Si](C)([O-])C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or lumps
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only