2-Chloro-5-fluorophenol, 98%, Thermo Scientific Chemicals
2-Chloro-5-fluorophenol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloro-5-fluorophenol, 98%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
5 g
Catalog number H28567.06
also known as H28567-06
Price (USD)/ Each
62.80
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
62.80
Add to cart
2-Chloro-5-fluorophenol, 98%, Thermo Scientific Chemicals
Catalog numberH28567.06
Price (USD)/ Each
62.80
-
Add to cart
Chemical Identifiers
CAS3827-49-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.5235-1.5275 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only