Thermo Scientific Chemicals

Vinylboronic anhydride pyridine complex, 95%, Thermo Scientific Chemicals

Catalog number: H28703.03
1 g, Each
Thermo Scientific Chemicals

Vinylboronic anhydride pyridine complex, 95%, Thermo Scientific Chemicals

Catalog number: H28703.03
1 g, Each
Quantity
Catalog number: H28703.03
also known as H28703-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
95010-17-6
IUPAC Name
pyridine; triethenyl-1,3,5,2,4,6-trioxatriborinane
Molecular Formula
C11H14B3NO3
InChI Key
YLHJACXHRQQNQR-UHFFFAOYSA-N
SMILES
C1=CC=NC=C1.C=CB1OB(OB(O1)C=C)C=C
Appearance (Color)
White to cream or pale yellow
Proton NMR
Conforms to structure
Assay by NMR
>94.0%
Form
Crystalline powder

Description

Reagent used for Suzuki-Miyaura cross-coupling, stereoselective synthesis via Palladium-catalyzed carboamination, Alkyl-connected 2-amino-6-vinylpurine (AVP) cross linking agent to cytosine base in RNA, Kaiser oxime resin-derived palladacycle as a recoverable polymeric precatalyst in Suzuki-Miyaura cross-coupling reactions in aqueous media and Kinetic resolution of phosphoryl and sulfonyl esters of binaphthol derivatives via Pd-catalyzed alcoholysis of their vinyl ethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent used for Suzuki-Miyaura cross-coupling, stereoselective synthesis via Palladium-catalyzed carboamination, Alkyl-connected 2-amino-6-vinylpurine (AVP) cross linking agent to cytosine base in RNA, Kaiser oxime resin-derived palladacycle as a recoverable polymeric precatalyst in Suzuki-Miyaura cross-coupling reactions in aqueous media and Kinetic resolution of phosphoryl and sulfonyl esters of binaphthol derivatives via Pd-catalyzed alcoholysis of their vinyl ethers.

Solubility
Soluble in methanol.

Notes
Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text