Thermo Scientific Chemicals

N-Methyl-1-naphthalenemethylamine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: H28719.09
10 g, Each
Thermo Scientific Chemicals

N-Methyl-1-naphthalenemethylamine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: H28719.09
10 g, Each
Quantity
Catalog number: H28719.09
also known as H28719-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
65473-13-4
IUPAC Name
methyl[(naphthalen-1-yl)methyl]azanium
Molecular Formula
C12H14N
InChI Key
MQRIUFVBEVFILS-UHFFFAOYSA-O
SMILES
C[NH2+]CC1=C2C=CC=CC2=CC=C1
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Appearance (Color)
White to cream
Melting Point (clear melt)
189-195?C
Form
Powder or crystalline powder
Assay (Freebase GC)
≥97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text