Thermo Scientific Chemicals

Methyl 3-iodo-4-methylbenzoate, 98%, Thermo Scientific Chemicals

Catalog number: H28734.14
25 g, Each
Thermo Scientific Chemicals

Methyl 3-iodo-4-methylbenzoate, 98%, Thermo Scientific Chemicals

Catalog number: H28734.14
25 g, Each
Quantity
Catalog number: H28734.14
also known as H28734-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
90347-66-3
IUPAC Name
methyl 3-iodo-4-methylbenzoate
Molecular Formula
C9H9IO2
InChI Key
NKMHAOTZPFVSPC-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC(I)=C(C)C=C1
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥97.5%
Refractive Index
1.5955-1.5995 @ 20?C
Form
Liquid

Description

Methyl 3-Iodo-4-methylbenzoate is used as a reagent in the synthesis of [(pyrazolo[1,5-a]pyridmidinyl)ethynyl]benzamides as selective discoidin domain receptor 1 (DDR1) inhibitors. Also used as a reagent in the preparation of orally bioavailable GZD824 targeting Breakpoint Cluster Region-Abelson (Bcr-Abl) kinase and overcoming antitumor drug resistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-Iodo-4-methylbenzoate is used as a reagent in the synthesis of [(pyrazolo[1,5-a]pyridmidinyl)ethynyl]benzamides as selective discoidin domain receptor 1 (DDR1) inhibitors. Also used as a reagent in the preparation of orally bioavailable GZD824 targeting Breakpoint Cluster Region-Abelson (Bcr-Abl) kinase and overcoming antitumor drug resistance.

Solubility
Not miscible or difficult to mix with water.

Notes
Light Sensitive, store in dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text