4-Methyl-2-(methylthio)pyrimidine, 98%, Thermo Scientific Chemicals
4-Methyl-2-(methylthio)pyrimidine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methyl-2-(methylthio)pyrimidine, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number H28795.06
also known as H28795-06
Price (USD)/ Each
69.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
69.00
Add to cart
4-Methyl-2-(methylthio)pyrimidine, 98%, Thermo Scientific Chemicals
Catalog numberH28795.06
Price (USD)/ Each
69.00
-
Add to cart
Chemical Identifiers
CAS14001-63-9
IUPAC Name4-methyl-2-(methylsulfanyl)pyrimidine
Molecular FormulaC6H8N2S
InChI KeyUCERVHYBSTYCQS-UHFFFAOYSA-N
SMILESCSC1=NC=CC(C)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to orange
Assay (GC)≥97.5%
Refractive Index1.5690-1.5730 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only