It is employed as a intermediates for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a intermediates for pharmaceutical.
Solubility
Soluble in methanol. Not miscible or difficult to mix with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
Josias B.G. Yaméogo et. al. Self-assembled biotransesterified cyclodextrins as potential Artemisinin nanocarriers. II: In vitro behavior toward the immune system and in vivo biodistribution assessment of unloaded nanoparticles. European Journal of Pharmaceutics and Biopharmaceutics, 1999, 88(3),
Xi Zhang; Fei Song; Maierhaba Taxipalati; Wei Wei; Fengqin Feng. Comparative study of surface-active properties and antimicrobial activities of disaccharide monoesters.PLoS ONE. 2014, 9(12), e114845.