2-Aminoacetophenone hydrochloride, 97%, Thermo Scientific Chemicals
2-Aminoacetophenone hydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Aminoacetophenone hydrochloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Catalog number H29117.09
also known as H29117-09
Price (USD)/ Each
230.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
230.00
Add to cart
2-Aminoacetophenone hydrochloride, 97%, Thermo Scientific Chemicals
Catalog numberH29117.09
Price (USD)/ Each
230.00
-
Add to cart
Chemical Identifiers
CAS5468-37-1
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White or pale yellow to brown
Assay (Titration ex Chloride)≥96.0% to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only