Thermo Scientific Chemicals

7-Methylxanthine, 98%, Thermo Scientific Chemicals

Catalog number: H29190.MD
250 mg, Each
Thermo Scientific Chemicals

7-Methylxanthine, 98%, Thermo Scientific Chemicals

Catalog number: H29190.MD
250 mg, Each
Quantity
Catalog number: H29190.MD
also known as H29190-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
552-62-5
IUPAC Name
7-methyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Molecular Formula
C6H6N4O2
InChI Key
PFWLFWPASULGAN-UHFFFAOYSA-N
SMILES
CN1C=NC2=C1C(=O)NC(=O)N2
Appearance (Color)
Pale cream to pale yellow or pale green
Form
Powder
Assay (HPLC)
≥97.5%

Description

It is used to metabolite theophylline and caffeine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to metabolite theophylline and caffeine.

Solubility
Soluble in dimethyl sulfoxide. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text