5-Hydroxy-2-nitrobenzoic acid, 99%, Thermo Scientific Chemicals
5-Hydroxy-2-nitrobenzoic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Hydroxy-2-nitrobenzoic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Catalog number H29194.09
also known as H29194-09
Price (USD)/ Each
273.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
273.00
Add to cart
5-Hydroxy-2-nitrobenzoic acid, 99%, Thermo Scientific Chemicals
Catalog numberH29194.09
Price (USD)/ Each
273.00
-
Add to cart
Chemical Identifiers
CAS610-37-7
IUPAC Name5-hydroxy-2-nitrobenzoic acid
Molecular FormulaC7H5NO5
InChI KeyBUHKQTKKZAXSMH-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(O)=CC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Melting Point (clear melt)168.0-174.0?C
Appearance (Color)Pale yellow
FormPowder
Assay (Silylated GC)≥98.5%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only