Thermo Scientific Chemicals

1-Boc-3-cyano-4-pyrrolidinone, 97%, Thermo Scientific Chemicals

Catalog number: H29306.06
5 g, Each
Thermo Scientific Chemicals

1-Boc-3-cyano-4-pyrrolidinone, 97%, Thermo Scientific Chemicals

Catalog number: H29306.06
5 g, Each
Quantity
Catalog number: H29306.06
also known as H29306-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
175463-32-8
IUPAC Name
tert-butyl 3-cyano-4-oxopyrrolidine-1-carboxylate
Molecular Formula
C10H14N2O3
InChI Key
RKUQLKRPWWYRIG-UHFFFAOYNA-N
SMILES
CC(C)(C)OC(=O)N1CC(C#N)C(=O)C1
Appearance (Color)
White to pale cream
Assay (GC)
≥96.0%
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text