3-Carboxy-5-nitrobenzeneboronic acid, 97%, Thermo Scientific Chemicals
3-Carboxy-5-nitrobenzeneboronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Carboxy-5-nitrobenzeneboronic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H29348.03
also known as H29348-03
Price (USD)/ Each
61.10
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
61.10
Add to cart
3-Carboxy-5-nitrobenzeneboronic acid, 97%, Thermo Scientific Chemicals
Catalog numberH29348.03
Price (USD)/ Each
61.10
-
Add to cart
Chemical Identifiers
CAS101084-81-5
IUPAC Name3-(dihydroxyboranyl)-5-nitrobenzoic acid
Molecular FormulaC7H6BNO6
InChI KeyWNIFCLWDGNHGMX-UHFFFAOYSA-N
SMILESOB(O)C1=CC(=CC(=C1)[N+]([O-])=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Melting Point (clear melt)225.0-235.0?C
FormPowder
Identification (FTIR)Conforms
Proton NMRConforms to structure
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only