Thermo Scientific Chemicals

Benzofurazan-5-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: H29358.MD
250 mg, Each
Thermo Scientific Chemicals

Benzofurazan-5-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: H29358.MD
250 mg, Each
Quantity
Catalog number: H29358.MD
also known as H29358-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1073355-14-2
IUPAC Name
5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,1,3-benzoxadiazole
Molecular Formula
C12H15BN2O3
InChI Key
MQXANAGMQAYTMV-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)C1=CC2=NON=C2C=C1
Form
Powder
Melting Point (clear melt)
84.5-90.5?C
Appearance (Color)
Cream
Assay (GC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text