Thermo Scientific Chemicals

1-Adamantanamine, 98%, Thermo Scientific Chemicals

Catalog number: H30076.06
5 g, Each
Thermo Scientific Chemicals

1-Adamantanamine, 98%, Thermo Scientific Chemicals

Catalog number: H30076.06
5 g, Each
Quantity
Catalog number: H30076.06
also known as H30076-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
768-94-5
IUPAC Name
adamantan-1-amine
Molecular Formula
C10H17N
InChI Key
DKNWSYNQZKUICI-UHFFFAOYSA-N
SMILES
NC12CC3CC(CC(C3)C1)C2
Form
Crystals or powder or crystalline powder.
Appearance (Color)
White
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Identification (FTIR)
Conforms

Description

1-Adamantanamine acts as a catalyst in enantioselective strecker reaction of phosphinoyl imines. It is also useful for making adamantane derivatives, i.e. amantadine hydrochloride. It is used in the pharmaceutical industry as antiviral and an antiparkinsonian drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantanamine acts as a catalyst in enantioselective strecker reaction of phosphinoyl imines. It is also useful for making adamantane derivatives, i.e. amantadine hydrochloride. It is used in the pharmaceutical industry as antiviral and an antiparkinsonian drug.

Solubility
Soluble in organic solvents. Insoluble in water.

Notes
Use only in a chemical fume hood. Keep away from sources of ignition. Incompatible with oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text