Thermo Scientific Chemicals

1,1'-Thiocarbonyldi-2(1H)-pyridone, 95%, Thermo Scientific Chemicals

Catalog number: H30087.06
5 g, Each
Thermo Scientific Chemicals

1,1'-Thiocarbonyldi-2(1H)-pyridone, 95%, Thermo Scientific Chemicals

Catalog number: H30087.06
5 g, Each
Quantity
Catalog number: H30087.06
also known as H30087-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
102368-13-8
IUPAC Name
1-(2-oxo-1,2-dihydropyridine-1-carbothioyl)-1,2-dihydropyridin-2-one
Molecular Formula
C11H8N2O2S
InChI Key
KXMMNJQMGILZDB-UHFFFAOYSA-N
SMILES
O=C1C=CC=CN1C(=S)N1C=CC=CC1=O
Appearance (Color)
Orange to red
Assay (HPLC)
≥94.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
153.0-166.0?C
Form
Powder

Description

1,1'-Thiocarbonyldi-2(1H)-pyridone is used in the preparation of thio-analogs of thioureas, sulforaphane and 2-furan-2-yl-3-hydroxy-6-isothiocyanato-chromen-4-one. It is involved in the preparation of norbiotinamine and its reactive derivatives such as isothiocyanates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1′-Thiocarbonyldi-2(1H)-pyridone is used in the preparation of thio-analogs of thioureas, sulforaphane and 2-furan-2-yl-3-hydroxy-6-isothiocyanato-chromen-4-one. It is involved in the preparation of norbiotinamine and its reactive derivatives such as isothiocyanates.

Notes
Incompatible with strong oxidizing agents. Store in a cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text