Thermo Scientific Chemicals

Benzophenone imine, 95%, Stab., Thermo Scientific Chemicals

Catalog number: H30110.06
5 g, Each
Thermo Scientific Chemicals

Benzophenone imine, 95%, Stab., Thermo Scientific Chemicals

Catalog number: H30110.06
5 g, Each
Quantity
Catalog number: H30110.06
also known as H30110-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1013-88-3
IUPAC Name
diphenylmethanimine
Molecular Formula
C13H11N
InChI Key
SXZIXHOMFPUIRK-UHFFFAOYSA-N
SMILES
N=C(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
Clear colorless to yellow to yellow-brown
Refractive Index
1.6150-1.6210 @ 20°C
Form
Liquid
Assay (GC)
≥94.0%
Stabilizer
Approx. 0.02% w/w Hydroquinone

Description

Useful in the preparation of nitrile ylide dimers.Benzophenone imine as ammonia surrogate. As intermediate material for synthesis of organic chemicals. Catalytic enantioselective alkylation of the benzophenone imine of glycine t-butyl ester using phosphazene (Schwesinger) bases and chiral quaternary ammonium salts yields either enantiomer of amino acid derivatives in excellent yield.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful in the preparation of nitrile ylide dimers.Benzophenone imine as ammonia surrogate. As intermediate material for synthesis of organic chemicals. Catalytic enantioselective alkylation of the benzophenone imine of glycine t-butyl ester using phosphazene (Schwesinger) bases and chiral quaternary ammonium salts yields either enantiomer of amino acid derivatives in excellent yield.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text