3,3-Dimethyl-1-butanol, 97%, Thermo Scientific Chemicals
3,3-Dimethyl-1-butanol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,3-Dimethyl-1-butanol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number H30194.18
also known as H30194-18
Price (USD)/ Each
374.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
374.00
Add to cart
3,3-Dimethyl-1-butanol, 97%, Thermo Scientific Chemicals
Catalog numberH30194.18
Price (USD)/ Each
374.00
-
Add to cart
Chemical Identifiers
CAS624-95-3
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4115-1.4165 @ 20?C
FormLiquid
Assay (GC)≥96.0%
Appearance (Color)Clear colorless
3,3-Dimethyl-1-butanol, is an organic building block used for the synthesis of various pharmaceutical compounds. It is an important intermediate in the synthesis of Neotame, an enhanced sweetening agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3-Dimethyl-1-butanol, is an organic building block used for the synthesis of various pharmaceutical compounds. It is an important intermediate in the synthesis of Neotame, an enhanced sweetening agent.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Setrak K. Tanielyan; Robert L. Augustine. Synthesis of 3,3-Dimethylbutanol and 3,3-Dimethylbutanal, Important Intermediates in the Synthesis of Neotame. Topics in Catalysis. 2012, 55(7), 625-630.
  2. R. E. Ranney; J. A. Oppermann; E. Muldoon; and F. G. McMahon. Comparative metabolism of aspartame in experimental animals and humans.Chinese Journal of Chromatography. 1976, 2(2), 441-451.