Thermo Scientific Chemicals

Methyl 3-bromo-4-hydroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: H30222.18
50 g, Each
Thermo Scientific Chemicals

Methyl 3-bromo-4-hydroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: H30222.18
50 g, Each
Quantity
Catalog number: H30222.18
also known as H30222-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
29415-97-2
IUPAC Name
methyl 3-bromo-4-hydroxybenzoate
Molecular Formula
C8H7BrO3
InChI Key
RKUNSPWAQIUGEZ-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC(Br)=C(O)C=C1
Appearance (Color)
White
Melting Point (clear melt)
104.0-110.0?C
Form
Powder
Assay (GC)
≥97.5%

Description

A reactant used in the preparation of selective inhibitors. In unsymmetrical Ullman reaction between methyl 5bromovanillate (II) and methyl 3-bromo-4-hydroxybenzoate (III) yielded all the three possible dicarbomethoxy-dibenzo-pdioxms which were separated by repeated chromatography over silica gel. A phosphorus- and bromine-containing bifunctional monomer has been prepared from bis(chloromethyl)methylphosphine oxide and the sodium salt of methyl 3-bromo-4-hydroxybenzoate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A reactant used in the preparation of selective inhibitors. In unsymmetrical Ullman reaction between methyl 5bromovanillate (II) and methyl 3-bromo-4-hydroxybenzoate (III) yielded all the three possible dicarbomethoxy-dibenzo-pdioxms which were separated by repeated chromatography over silica gel. A phosphorus- and bromine-containing bifunctional monomer has been prepared from bis(chloromethyl)methylphosphine oxide and the sodium salt of methyl 3-bromo-4-hydroxybenzoate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text