Thermo Scientific Chemicals

Carbon black, Super P∣r Conductive, 99+% (metals basis), Thermo Scientific Chemicals

Catalog number: H30253.22
100 g, Each
Thermo Scientific Chemicals

Carbon black, Super P∣r Conductive, 99+% (metals basis), Thermo Scientific Chemicals

Catalog number: H30253.22
100 g, Each
Quantity
Catalog number: H30253.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1333-86-4
IUPAC Name
carbon
Molecular Formula
C
InChI Key
OKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES
[C]
Sulfated Ash
≤0.100%
Surface Area
BET Surface Area: 57.0-67.0 m²/g
Appearance (Color)
Black
Form
Powder
Comment
Absorption Stiffness Value: 30.0-34.0 ml/5g

Description

Carbon black Super-P (TIMCAL) was used as conductive agent. Super P furnace black the best conductive additive. Carbon black (conducting material, super P black) was added with binder in the composite electrode to compensate the low electrical conductivity of PPy and PPyDVB in miniemulsion polymerization. The hybrid Super P-SACNT conductive network manifests itself as a promising strategy to improve the battery performances with a minimum amount of conductive fillers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carbon black Super-P (TIMCAL) was used as conductive agent. Super P furnace black the best conductive additive. Carbon black (conducting material, super P black) was added with binder in the composite electrode to compensate the low electrical conductivity of PPy and PPyDVB in miniemulsion polymerization. The hybrid Super P-SACNT conductive network manifests itself as a promising strategy to improve the battery performances with a minimum amount of conductive fillers.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text