Thermo Scientific Chemicals

3-Hydroxybutyric acid, 97%, Thermo Scientific Chemicals

Catalog number: H30426.06
5 g, Each
Thermo Scientific Chemicals

3-Hydroxybutyric acid, 97%, Thermo Scientific Chemicals

Catalog number: H30426.06
5 g, Each
Quantity
Catalog number: H30426.06
also known as H30426-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
300-85-6
IUPAC Name
3-hydroxybutanoic acid
Molecular Formula
C4H8O3
InChI Key
WHBMMWSBFZVSSR-UHFFFAOYNA-N
SMILES
CC(O)CC(O)=O
Form
Crystals or powder or crystalline powder or fused or granular solid or lumps
Water Content (Karl Fischer Titration)
≤0.75%
Assay (Aqueous acid-base Titration)
≥96.0% to ≤104.0%
Assay (Silylated GC)
≥96.0%
Appearance (Color)
White to pale cream

Description

3-Hydroxybutyric acid is used in enantioselective synthesis of D-(-)-3-hydroxybutyric acid by escherichia coli. It forms films of random copolymer with (R)-3-hydroxypentanoic acid, (R)-3-hydroxyhexanoic acid, 4-hydroxybutyric acid, 6-hydroxyhexanoic acid and (S,S)-lactide by the melt-crystallized method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxybutyric acid is used in enantioselective synthesis of D-(-)-3-hydroxybutyric acid by escherichia coli. It forms films of random copolymer with (R)-3-hydroxypentanoic acid, (R)-3-hydroxyhexanoic acid, 4-hydroxybutyric acid, 6-hydroxyhexanoic acid and (S,S)-lactide by the melt-crystallized method.

Solubility
Miscible with water, alcohol and ether.

Notes
Hygroscopic. Incompatible with acids and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text