3-Hydroxybutyric acid, 97%, Thermo Scientific Chemicals
3-Hydroxybutyric acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Hydroxybutyric acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number H30426.14
also known as H30426-14
Price (USD)/ Each
638.65
Online exclusive
709.00 
Save 70.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
638.65
Online exclusive
709.00 
Save 70.35 (10%)
Add to cart
3-Hydroxybutyric acid, 97%, Thermo Scientific Chemicals
Catalog numberH30426.14
Price (USD)/ Each
638.65
Online exclusive
709.00 
Save 70.35 (10%)
-
Add to cart
Chemical Identifiers
CAS300-85-6
IUPAC Name3-hydroxybutanoic acid
Molecular FormulaC4H8O3
InChI KeyWHBMMWSBFZVSSR-UHFFFAOYNA-N
SMILESCC(O)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥96.0% to ≤104.0%
FormCrystals or powder or crystalline powder or fused or granular solid or lumps
Water Content (Karl Fischer Titration)≤0.75%
Assay (Silylated GC)≥96.0%
Appearance (Color)White to pale cream
3-Hydroxybutyric acid is used in enantioselective synthesis of D-(-)-3-hydroxybutyric acid by escherichia coli. It forms films of random copolymer with (R)-3-hydroxypentanoic acid, (R)-3-hydroxyhexanoic acid, 4-hydroxybutyric acid, 6-hydroxyhexanoic acid and (S,S)-lactide by the melt-crystallized method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxybutyric acid is used in enantioselective synthesis of D-(-)-3-hydroxybutyric acid by escherichia coli. It forms films of random copolymer with (R)-3-hydroxypentanoic acid, (R)-3-hydroxyhexanoic acid, 4-hydroxybutyric acid, 6-hydroxyhexanoic acid and (S,S)-lactide by the melt-crystallized method.

Solubility
Miscible with water, alcohol and ether.

Notes
Hygroscopic. Incompatible with acids and bases.
RUO – Research Use Only

General References:

  1. Bredikhin, A. A.; Bredikhina, Z. A.; Zakharychev, D. V.; Samigullina, A. I.; Gubaidullin, A. T. 4-Benzoylamino-3-Hydroxybutyric acid, Historically First Anomalous Racemate: Reinvestigation. Crys. Growth Des. 2015, 15 (3), 1362-1373.
  2. Numata, K.; Morisaki, K. Screening of Marine Bacteria To Synthesize Polyhydroxyalkanoate from Lignin: Contribution of Lignin Derivatives to Biosynthesis by Oceanimonas doudoroffii. ACS Sustainable Chem. Eng. 2015, 3 (4), 569-573.