4-Ethylbenzyl alcohol, 99%, Thermo Scientific Chemicals
4-Ethylbenzyl alcohol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Ethylbenzyl alcohol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H30459.06
also known as H30459-06
Price (USD)/ Each
156.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
156.00
Add to cart
4-Ethylbenzyl alcohol, 99%, Thermo Scientific Chemicals
Catalog numberH30459.06
Price (USD)/ Each
156.00
-
Add to cart
Chemical Identifiers
CAS768-59-2
IUPAC Name(4-ethylphenyl)methanol
Molecular FormulaC9H12O
InChI KeyYSLBFFIVJGJBSA-UHFFFAOYSA-N
SMILESCCC1=CC=C(CO)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.5%
Refractive Index1.5260-1.5290 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only