6-Bromopyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals
6-Bromopyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Bromopyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H30553.06
also known as H30553-06
Price (USD)/ Each
163.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
163.00
Add to cart
6-Bromopyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals
Catalog numberH30553.06
Price (USD)/ Each
163.00
-
Add to cart
Chemical Identifiers
CAS21190-87-4
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)> 96.0%
Formpowder
Sulfated Ash< 1.50%
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)> 96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only