Thermo Scientific Chemicals

Thieno[3,2-b]thiophene, 97%, Thermo Scientific Chemicals

Catalog number: H30637.06
5 g, Each
Thermo Scientific Chemicals

Thieno[3,2-b]thiophene, 97%, Thermo Scientific Chemicals

Catalog number: H30637.06
5 g, Each
Quantity
Catalog number: H30637.06
also known as H30637-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
251-41-2
IUPAC Name
thieno[3,2-b]thiophene
Molecular Formula
C6H4S2
InChI Key
VJYJJHQEVLEOFL-UHFFFAOYSA-N
SMILES
S1C=CC2=C1C=CS2
Identification (FTIR)
Conforms
Appearance (Color)
White to cream to brown to gray
Assay (GC)
≥96.0%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
51.0-60.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text