Thermo Scientific Chemicals

tert-Butyl vinyl ether, 98%, stab. with ca 0.1% N,N-diethylaniline, Thermo Scientific Chemicals

Catalog number: H30770.06
5 g, Each
Thermo Scientific Chemicals

tert-Butyl vinyl ether, 98%, stab. with ca 0.1% N,N-diethylaniline, Thermo Scientific Chemicals

Catalog number: H30770.06
5 g, Each
Quantity
Catalog number: H30770.06
also known as H30770-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
926-02-3
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥97.5%
Refractive Index
1.3965-1.4005 @ 20?C

Description

tert-Butyl vinyl ether is used in the synthesis of 3-tert-Butoxycyclobutanone by reacting with ketene using zinc(II) chloride as a catalyst. It acts as an intermediate for the functional vinyl monomer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl vinyl ether is used in the synthesis of 3-tert-Butoxycyclobutanone by reacting with ketene using zinc(II) chloride as a catalyst. It acts as an intermediate for the functional vinyl monomer.

Solubility
Miscible with alcohol, carbon disulfide, chloroform and diethyl ether. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text