Thermo Scientific Chemicals

3-Methyl-4-nitro-1H-pyrazole, 97%, Thermo Scientific Chemicals

Catalog number: H30860.14
25 g, Each
Thermo Scientific Chemicals

3-Methyl-4-nitro-1H-pyrazole, 97%, Thermo Scientific Chemicals

Catalog number: H30860.14
25 g, Each
Quantity
Catalog number: H30860.14
also known as H30860-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5334-39-4
IUPAC Name
5-methyl-4-nitro-1H-pyrazole
Molecular Formula
C4H5N3O2
InChI Key
WTZYTQJELOHMMJ-UHFFFAOYSA-N
SMILES
CC1=C(C=NN1)[N+]([O-])=O
Melting Point (clear melt)
129.0-139.0?C
Appearance (Color)
White to pale cream to cream to yellow
Assay (GC)
≥96.0%
Form
Crystals or powder or crystalline powder

Description

3-Methyl-4-nitropyrazole is a reactant used in the preparation of aminopyrazole derivatives as potent, selective and brain penetrant Leucine-rich repeat kinase 2(LRRK2) small molecule inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-4-nitropyrazole is a reactant used in the preparation of aminopyrazole derivatives as potent, selective and brain penetrant Leucine-rich repeat kinase 2(LRRK2) small molecule inhibitors.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text