Thermo Scientific Chemicals

3-Amino-1-methyl-1H-pyrazole, 97%, Thermo Scientific Chemicals

Catalog number: H30935.06
5 g, Each
Thermo Scientific Chemicals

3-Amino-1-methyl-1H-pyrazole, 97%, Thermo Scientific Chemicals

Catalog number: H30935.06
5 g, Each
Quantity
Catalog number: H30935.06
also known as H30935-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1904-31-0
IUPAC Name
1-methyl-1H-pyrazol-3-amine
Molecular Formula
C4H7N3
InChI Key
MOGQNVSKBCVIPW-UHFFFAOYSA-N
SMILES
CN1C=CC(N)=N1
Appearance (Color)
Clear colorless to yellow or pale orange to red
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.5395-1.5445 @ 20°C

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text