Thermo Scientific Chemicals

Bromomalonaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: H31030.18
50 g, Each
Thermo Scientific Chemicals

Bromomalonaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: H31030.18
50 g, Each
Quantity
Catalog number: H31030.18
also known as H31030-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2065-75-0
IUPAC Name
2-bromopropanedial
Molecular Formula
C3H3BrO2
InChI Key
SURMYNZXHKLDFO-UHFFFAOYSA-N
SMILES
BrC(C=O)C=O
Assay (Aqueous acid-base Titration)
≥96.0%
Appearance (Color)
Pale yellow to brown
Form
Crystals or powder or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text