Thermo Scientific Chemicals

trans-4-Aminocyclohexanecarboxylic acid hydrochloride, 96%, Thermo Scientific Chemicals

Catalog number: H31114.06
5 g, Each
Thermo Scientific Chemicals

trans-4-Aminocyclohexanecarboxylic acid hydrochloride, 96%, Thermo Scientific Chemicals

Catalog number: H31114.06
5 g, Each
Quantity
Catalog number: H31114.06
also known as H31114-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
27960-59-4
IUPAC Name
4-aminocyclohexane-1-carboxylic acid hydrochloride
Molecular Formula
C7H14ClNO2
InChI Key
HXZSYUOXTKQNNY-UHFFFAOYSA-N
SMILES
Cl.NC1CCC(CC1)C(O)=O
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥95.0 to ≤105.0%
Water Content (Karl Fischer Titration)
≤2.0%
Assay (Titration ex Chloride)
≥95.0 to ≤105.0%

Description

Used as intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as intermediate in organic synthesis.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text