Chemical Identifiers
CAS84783-64-2
IUPAC Name[2'-(diphenylphosphanyl)-[1,1'-biphenyl]-2-yl]diphenylphosphane
Molecular FormulaC36H28P2
InChI KeyGRTJBNJOHNTQBO-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
View more