Acetoxyacetone, 97%, Thermo Scientific Chemicals
Acetoxyacetone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetoxyacetone, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
50 mL
Catalog number H31346.AB
also known as H31346-AB
Price (USD)/ Each
85.65
Online exclusive
95.30 
Save 9.65 (10%)
-
Add to cart
Quantity:
10 mL
Request bulk or custom format
Price (USD)/ Each
85.65
Online exclusive
95.30 
Save 9.65 (10%)
Add to cart
Acetoxyacetone, 97%, Thermo Scientific Chemicals
Catalog numberH31346.AB
Price (USD)/ Each
85.65
Online exclusive
95.30 
Save 9.65 (10%)
-
Add to cart
Chemical Identifiers
CAS592-20-1
IUPAC Name2-oxopropyl acetate
Molecular FormulaC5H8O3
InChI KeyDBERHVIZRVGDFO-UHFFFAOYSA-N
SMILESCC(=O)COC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4130-1.4180 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Acetoxyacetone is an building block in the synthesis of various pharmaceutical and organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetoxyacetone is an building block in the synthesis of various pharmaceutical and organic compounds.

Solubility
Soluble in diethyl ether, ethyl acetate and dichloromethane.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. W.C. Sung. Volatile constituents detected in smoke condensates from the combination of the smoking ingredients sucrose, black tea leaves, and bread flour. 2013, 21292-300.
  2. John Morrow Stewart; D. W. Woolley. Antimetabolites of Mevalonic Acid. J. Am. Chem. Soc,1959, 81(18), 4951-4956.