Ethyl 2-cyano-2-methylpropionate, 97%, Thermo Scientific Chemicals
Ethyl 2-cyano-2-methylpropionate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 2-cyano-2-methylpropionate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number H31432.18
also known as H31432-18
Price (USD)/ Each
282.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
282.00
Add to cart
Ethyl 2-cyano-2-methylpropionate, 97%, Thermo Scientific Chemicals
Catalog numberH31432.18
Price (USD)/ Each
282.00
-
Add to cart
Chemical Identifiers
CAS1572-98-1
IUPAC Nameethyl 2-cyano-2,2-dimethylacetate
Molecular FormulaC7H11NO2
InChI KeyFYGRPGOHQCPZCV-UHFFFAOYSA-N
SMILESCCOC(=O)C(C)(C)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Refractive Index1.4075-1.4125 @ 20?C
FormLiquid
Assay (GC)≥96.0%
It is employed as a intermediates for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediates for pharmaceutical and organic synthesis.

Solubility
Soluble in ether. Not miscible or difficult to mix with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Hugh W. Thompson; Joseph Swistok. Enamines of 3,3-dimethylazetidine. J. Org. Chem,1981, 46(24), 4907-4911.
  2. Ulhas P. Dhokte; Vien V. Khau; Darrell R. Hutchison; Michael J. Martinelli. A novel approach for total synthesis of cryptophycins via asymmetric crotylboration protocol. Tetrahedron Letters,1998, 39(48), 8771-8774.