Ethyl thiooxamate, 95%, Thermo Scientific Chemicals
Ethyl thiooxamate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl thiooxamate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number H31447.06
also known as H31447-06
Price (USD)/ Each
137.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
137.00
Add to cart
Ethyl thiooxamate, 95%, Thermo Scientific Chemicals
Catalog numberH31447.06
Price (USD)/ Each
137.00
-
Add to cart
Chemical Identifiers
CAS16982-21-1
IUPAC Nameethyl carbamothioylformate
Molecular FormulaC4H7NO2S
InChI KeyYMBMCMOZIGSBOA-UHFFFAOYSA-N
SMILESCCOC(=O)C(N)=S
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥94.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)55.0-68.0?C
Appearance (Color)Yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only