Thermo Scientific Chemicals

4,4'-Di-n-nonyl-2,2'-bipyridine, 97%, Thermo Scientific Chemicals

Catalog number: H31476.06
5 g, Each
Thermo Scientific Chemicals

4,4'-Di-n-nonyl-2,2'-bipyridine, 97%, Thermo Scientific Chemicals

Catalog number: H31476.06
5 g, Each
Quantity
Catalog number: H31476.06
also known as H31476-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
142646-58-0
IUPAC Name
4,4'-dinonyl-2,2'-bipyridine
Molecular Formula
C28H44N2
InChI Key
VHJFWJXYEWHCGD-UHFFFAOYSA-N
SMILES
CCCCCCCCCC1=CC(=NC=C1)C1=NC=CC(CCCCCCCCC)=C1
Assay (Non-aqueous acid-base Titration)
≥96.0 to ≤104.0%
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
57.0-67.0?C

Description

Used in the preparation of an artificial muscle with lamellar structure based on a nematic triblock copolymer. Also use as complex in cyclic voltammetric studies of copper complexes catalyzing atom transfer radical polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the preparation of an artificial muscle with lamellar structure based on a nematic triblock copolymer. Also use as complex in cyclic voltammetric studies of copper complexes catalyzing atom transfer radical polymerization.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text