4-(Trifluoromethylthio)benzylamine, 95%, Thermo Scientific Chemicals
4-(Trifluoromethylthio)benzylamine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(Trifluoromethylthio)benzylamine, 95%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number H31515.03
also known as H31515-03
Price (USD)/ Each
131.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
131.00
Add to cart
4-(Trifluoromethylthio)benzylamine, 95%, Thermo Scientific Chemicals
Catalog numberH31515.03
Price (USD)/ Each
131.00
-
Add to cart
Chemical Identifiers
CAS128273-56-3
IUPAC Name1-{4-[(trifluoromethyl)sulfanyl]phenyl}methanamine
Molecular FormulaC8H8F3NS
InChI KeyLACURGWEZCFLBO-UHFFFAOYSA-N
SMILESNCC1=CC=C(SC(F)(F)F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5005-1.5065 @ 20?C
FormLiquid
Assay (GC)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only