2,5-Difluoro-4-hydroxybenzaldehyde is used as primary and secondary intermediate and as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,5-Difluoro-4-hydroxybenzaldehyde is used as primary and secondary intermediate and as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Store away from oxidizing agents, air. Store under inert gas.
RUO – Research Use Only
Carla I. Nieto; M. Pilar Cabildo; M. Pilar Cornago; Dionisia Sanz; Rosa M. Claramunt; Ibon Alkorta; José Elguero; José A. García; Ana López; Darío Acuña-Castroviejo. Synthesis, structure and biological activity of 3(5)-trifluoromethyl-1H-pyrazoles derived from hemicurcuminoids. Journal of Molecular Structure. 2003, 1100, 518-529.