Thermo Scientific Chemicals

1,1'-Diethyl-2,2'-carbocyanine iodide, 96%, Thermo Scientific Chemicals

Catalog number: H31540.MD
250 mg, Each
Thermo Scientific Chemicals

1,1'-Diethyl-2,2'-carbocyanine iodide, 96%, Thermo Scientific Chemicals

Catalog number: H31540.MD
250 mg, Each
Quantity
Catalog number: H31540.MD
also known as H31540-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
605-91-4
IUPAC Name
1-ethyl-2-[3-(1-ethyl-1,2-dihydroquinolin-2-ylidene)prop-1-en-1-yl]quinolin-1-ium iodide
Molecular Formula
C25H25IN2
InChI Key
QWYZFXLSWMXLDM-UHFFFAOYSA-M
SMILES
[I-].CCN1C(=CC=CC2=CC=C3C=CC=CC3=[N+]2CC)C=CC2=CC=CC=C12
Elemental Analysis
I: 25.10-27.74% (Theory: 26.42%)
Appearance (Color)
Dark green
Form
Powder
Elemental Analysis
N: 5.54-6.12% (Theory: 5.83%)

Description

The fluorescence decay time of pinacyanol (1,1?-diethyl-2,2?-carbocyanine) iodide has been measured in the temperature range 95-225 K.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The fluorescence decay time of pinacyanol (1,1′-diethyl-2,2′-carbocyanine) iodide has been measured in the temperature range 95-225 K.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text