Thermo Scientific Chemicals

4-Fluoro-2,3-dimethylaniline, 97%, Thermo Scientific Chemicals

Catalog number: H31560.03
1 g, Each
Thermo Scientific Chemicals

4-Fluoro-2,3-dimethylaniline, 97%, Thermo Scientific Chemicals

Catalog number: H31560.03
1 g, Each
Quantity
Catalog number: H31560.03
also known as H31560-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1737-68-4
IUPAC Name
4-fluoro-2,3-dimethylaniline
Molecular Formula
C8H10FN
InChI Key
LKMJLYCMLAPNDB-UHFFFAOYSA-N
SMILES
CC1=C(N)C=CC(F)=C1C
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
38.0-48.0?C
Appearance (Color)
White to pale pink
Assay (GC)
≥96.0%

Description

4-Fluoro-2,3-dimethylaniline is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-2,3-dimethylaniline is used as a pharmaceutical intermediate.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text