4-Hydroxybenzamidine hydrochloride is used as a pharmaceutical intermediate and used in organic chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Hydroxybenzamidine hydrochloride is used as a pharmaceutical intermediate and used in organic chemical synthesis.
Solubility
Soluble in water.
Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture.
RUO – Research Use Only
A. Boller; M. Cereghetti; M. Schadt & H. Scherrer. Synthesis and some Physical Properties of Phenylpyrimidines. Molecular Crystals and Liquid Crystals. 1977, 71 (18), 215-231.
Jean Jacques Vanden Eynde; Annie Mayence; Tien L Huang; Margaret S Collins; Sandra Rebholz; Peter D Walzer; Melanie T Cushion. Novel bisbenzamidines as potential drug candidates for the treatment of Pneumocystis carinii pneumonia. Bioorganic & Medicinal Chemistry Letters. 2004, 14 (17), 4545-4548.