Thermo Scientific Chemicals

Hydrogen chloride, ca 0.5M soln. in methanol, Thermo Scientific Chemicals

Catalog number: H31570.AE
100 mL, Each
Thermo Scientific Chemicals

Hydrogen chloride, ca 0.5M soln. in methanol, Thermo Scientific Chemicals

Catalog number: H31570.AE
100 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: H31570.AE
also known as H31570-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7647-01-0
IUPAC Name
hydrogen chloride
Molecular Formula
ClH
InChI Key
VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES
Cl
Form
Liquid
Appearance (Color)
Clear, colorless
Assay (Aqueous acid-base Titration)
0.4 - 0.6M

Description

Industrially used in the production of inorganic salts of vinyl chloride from acetylene, alkyl chlorides from olefins, dyes, artificial silk, pigment, pharmaceutical hydrochloride, Also as catalyst and solvent in organic syntheses, Alcohol chlorination reagent, Nitration reaction, Manufacture of phosphoric acid and in the production of ammonium chloride, Neutralization of basic systems, Cleaning agents. Manufacture of fertilizers, Hydrolysing of starch and proteins in the preparation of various food products, Refining ore Leather deliming / tanning agents, Oil- and gas-well treatment, Textile scouring agents, Removing scale from boilers and heat-exchange equipment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text