Thermo Scientific Chemicals

2,3,4,6-Tetra-O-benzyl-alpha-D-glucopyranose, 98%

Catalog number: H31595.03
1 g, Each
Thermo Scientific Chemicals

2,3,4,6-Tetra-O-benzyl-alpha-D-glucopyranose, 98%

Catalog number: H31595.03
1 g, Each
Quantity
Catalog number: H31595.03
also known as H31595-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6564-72-3
IUPAC Name
3,4,5-tris(benzyloxy)-6-[(benzyloxy)methyl]oxan-2-ol
Molecular Formula
C34H36O6
InChI Key
OGOMAWHSXRDAKZ-UHFFFAOYNA-N
SMILES
OC1OC(COCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C1OCC1=CC=CC=C1
Form
Powder
Appearance (Color)
White
Optical Rotation
49.8 ± 0.1? (C=2 in dioxane)
Melting Point (clear melt)
149.0-156.0?C

Description

It is the intermediate of Voglibose/Dapagliflozin. An important D-glucopyranose derivative for glucosylations and other reactions 1,2; Preparation of the α-glucopyranosyl chloride, synthesis of 1-C-α-D-glucopyranose derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is the intermediate of Voglibose/Dapagliflozin. An important D-glucopyranose derivative for glucosylations and other reactions 1,2; Preparation of the α-glucopyranosyl chloride, synthesis of 1-C-α-D-glucopyranose derivatives.

Solubility
Soluble in CHCl3. Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text